Merck
  • Important Role of the GLP-1 Axis for Glucose Homeostasis after Bariatric Surgery.
Important Role of the GLP-1 Axis for Glucose Homeostasis after Bariatric Surgery.
Cell reports (2019-02-07)
Pierre Larraufie, Geoffrey P Roberts, Anne K McGavigan, Richard G Kay, Joyce Li, Andrew Leiter, Audrey Melvin, Emma K Biggs, Peter Ravn, Kathleen Davy, David C Hornigold, Giles S H Yeo, Richard H Hardwick, Frank Reimann, Fiona M Gribble
ABSTRACT
Bariatric surgery is widely used to treat obesity and improves type 2 diabetes beyond expectations from the degree of weight loss. Elevated post-prandial concentrations of glucagon-like peptide 1 (GLP-1), peptide YY (PYY), and insulin are widely reported, but the importance of GLP-1 in post-bariatric physiology remains debated. Here, we show that GLP-1 is a major driver of insulin secretion after bariatric surgery, as demonstrated by blocking GLP-1 receptors (GLP1Rs) post-gastrectomy in lean humans using Exendin-9 or in mice using an anti-GLP1R antibody. Transcriptomics and peptidomics analyses revealed that human and mouse enteroendocrine cells were unaltered post-surgery; instead, we found that elevated plasma GLP-1 and PYY correlated with increased nutrient delivery to the distal gut in mice. We conclude that increased GLP-1 secretion after bariatric surgery arises from rapid nutrient delivery to the distal gut and is a key driver of enhanced insulin secretion.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O