Merck
  • Measurement of human surfactant protein-B turnover in vivo from tracheal aspirates using targeted proteomics.
Measurement of human surfactant protein-B turnover in vivo from tracheal aspirates using targeted proteomics.
Analytical chemistry (2010-02-25)
Daniela M Tomazela, Bruce W Patterson, Elizabeth Hanson, Kimberly L Spence, Tiffany B Kanion, David H Salinger, Paolo Vicini, Hugh Barret, Hillary B Heins, F Sessions Cole, Aaron Hamvas, Michael J MacCoss
ABSTRACT
We describe a method to measure protein synthesis and catabolism in humans without prior purification and use the method to measure the turnover of surfactant protein-B (SP-B). SP-B, a lung-specific, hydrophobic protein essential for fetal-neonatal respiratory transition, is present in only picomolar quantities in tracheal aspirate samples and difficult to isolate for dynamic turnover studies using traditional in vivo tracer techniques. Using infusion of [5,5,5-(2)H(3)] leucine and a targeted proteomics method, we measured both the quantity and kinetics of SP-B tryptic peptides in tracheal aspirate samples of symptomatic newborn infants. The fractional synthetic rate (FSR) of SP-B measured using the most abundant proteolytic fragment, a 10 amino acid peptide from the carboxy-terminus of proSP-B (SPTGEWLPR), from the circulating leucine pool was 0.035 +/- 0.005 h(-1), and the fractional catabolic rate was 0.044 +/- 0.003 h(-1). This technique permits high-throughput and sensitive measurement of turnover of low abundance proteins with minimal sample preparation.