Merck
  • Protein arginine methylation modulates light-harvesting antenna translation in Chlamydomonas reinhardtii.
Protein arginine methylation modulates light-harvesting antenna translation in Chlamydomonas reinhardtii.
The Plant journal : for cell and molecular biology (2010-12-24)
Olga Blifernez, Lutz Wobbe, Karsten Niehaus, Olaf Kruse
ABSTRACT
Methylation of protein arginines represents an important post-translational modification mechanism, which has so far primarily been characterized in mammalian cells. In this work, we successfully identified and characterized arginine methylation as a crucial type of post-translational modification in the activity regulation of the cytosolic translation repressor protein NAB1 in the plant model organism Chlamydomonas reinhardtii. NAB1 represses the cytosolic translation of light-harvesting protein encoding mRNAs by sequestration into translationally silent messenger ribonucleoprotein complexes (mRNPs). Protein arginine methylation of NAB1 could be demonstrated by PRMT1 catalyzed methylation of recombinant NAB1 in vitro, and by immunodetection of methylated NAB1 arginines in vivo. Mass spectrometric analyses of NAB1 purified from C. reinhardtii revealed the asymmetric dimethylation of Arg90 and Arg92 within GAR motif I. Inhibition of arginine methylation by either adenosine-2'-3'-dialdehyde (AdOx) or 7,7'-carbonylbis(azanediyl)bis(4-hydroxynaphthalene-2-sulfonic acid) sodium salt hydrate (AMI-1) caused a dark-green phenotype characterized by the increased accumulation of light-harvesting complex proteins, and indicating a reduced translation repressor activity of NAB1. The extent of NAB1 arginine methylation depends on the growth conditions, with phototrophic growth causing a high methylation state and heterotrophic growth resulting in lowered methylation of the protein. In addition, we could show that NAB1 activity regulation by arginine methylation operates independently from cysteine-based redox control, which has previously been shown to control the activity of NAB1.
Sigma-Aldrich
Adenosine, periodate oxidized, ≥93%
Sigma-Aldrich
AMI-1 sodium salt hydrate, ≥98% (HPLC)