Merck
Synthesis of carbon-14 labeled ZJ0712, a novel strobilurin fungicide.
Journal of labelled compounds & radiopharmaceuticals (2018-06-14)
Zheng-Min Yang, Ya-Jun Xu, Gui-Hua Zhang, Cai-Guang Jian, Xiao Li, Qing-Fu Ye
ABSTRACT
ZJ0712, a broad-spectrum fungicidal ingredient of strobilurin, exhibits a high protective and curative activity against plant pathogenic fungi. To support the study on its metabolism, residue, environmental behavior, and fate for safety evaluation, two versions of carbon-14 labeled ZJ0712, methyl (E)-2-(2-((2,5-dimethylphenoxy)methyl)phenyl)-3-methoxy[2-14 C]acrylate (2) and methyl (E)-2-(2-((2,5-dimethyl[phenyl-U-14 C6 ]phenoxy)methyl)phenyl)-3-methoxyacrylate (3), were synthesized from barium [14 C]carbonate in 6-step yield of 47% and from 2,5-dimethyl[phenyl-U-14 C6 ]phenol in the yield of 91%, respectively.
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, suitable for scintillation, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)