Merck
  • Secreted Frizzled-Related Protein-2 Inhibits Doxorubicin-Induced Apoptosis Mediated through the Akt-mTOR Pathway in Soleus Muscle.
Secreted Frizzled-Related Protein-2 Inhibits Doxorubicin-Induced Apoptosis Mediated through the Akt-mTOR Pathway in Soleus Muscle.
Oxidative medicine and cellular longevity (2018-08-29)
Hilda Merino, Dinender K Singla
ABSTRACT
Doxorubicin (Dox) is a potent chemotherapeutic drug known for its dose-dependent and serious adverse effects, such as cardiotoxicity and myotoxicity. Dox-induced cardiotoxicity (DIC) and muscle toxicity (DIMT) have been studied; however, the mechanisms of Dox-induced apoptosis in soleus muscle are not well defined. Our data shows that with Dox treatment, there is a significant increase in oxidative stress, apoptosis, proapoptotic protein BAX, pPTEN levels, and wnt3a and β-catenin activity (p < 0.05). Moreover, Dox treatment also resulted in decreased antioxidant levels, antiapoptotic BCL2, pAKT, p-mTOR, and endogenous levels of sFRP2 in the soleus muscle tissue (p < 0.05). Secreted frizzled-related protein 2 (sFRP2) treatment attenuated the adverse effects of DIMT and apoptosis in the soleus muscle, evidenced by a decrease in oxidative stress, apoptosis, BAX, pPTEN, and wnt3a and β-catenin activity, as well as an increase in antioxidants, BCL2, pAKT, p-MTOR, and sFRP2 levels (p < 0.05). This data suggests that Dox-induced oxidative stress and apoptosis is mediated through both the Akt-mTOR and wnt/β-catenin pathways. Moreover, the data also shows that sFRP2 modulates these two pathways by increasing signaling of Akt-mTOR and decreased signaling of the wnt/β-catenin pathway. Therefore, our data suggests that sFRP2 has valuable therapeutic potential in reversing Dox-induced oxidative stress and apoptosis in soleus muscle mediated through the Akt-mTOR pathway.
Sigma-Aldrich
Anti-Myosin (Skeletal) antibody produced in rabbit, whole antiserum
SKU
Pack Size
Availability
Price
Quantity
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests