Merck
  • Targeting PFKFB3 radiosensitizes cancer cells and suppresses homologous recombination.
Targeting PFKFB3 radiosensitizes cancer cells and suppresses homologous recombination.
Nature communications (2018-09-27)
Nina M S Gustafsson, Katarina Färnegårdh, Nadilly Bonagas, Anna Huguet Ninou, Petra Groth, Elisee Wiita, Mattias Jönsson, Kenth Hallberg, Jemina Lehto, Rosa Pennisi, Jessica Martinsson, Carina Norström, Jessica Hollers, Johan Schultz, Martin Andersson, Natalia Markova, Petra Marttila, Baek Kim, Martin Norin, Thomas Olin, Thomas Helleday
ABSTRACT
The glycolytic PFKFB3 enzyme is widely overexpressed in cancer cells and an emerging anti-cancer target. Here, we identify PFKFB3 as a critical factor in homologous recombination (HR) repair of DNA double-strand breaks. PFKFB3 rapidly relocates into ionizing radiation (IR)-induced nuclear foci in an MRN-ATM-γH2AX-MDC1-dependent manner and co-localizes with DNA damage and HR repair proteins. PFKFB3 relocalization is critical for recruitment of HR proteins, HR activity, and cell survival upon IR. We develop KAN0438757, a small molecule inhibitor that potently targets PFKFB3. Pharmacological PFKFB3 inhibition impairs recruitment of ribonucleotide reductase M2 and deoxynucleotide incorporation upon DNA repair, and reduces dNTP levels. Importantly, KAN0438757 induces radiosensitization in transformed cells while leaving non-transformed cells unaffected. In summary, we identify a key role for PFKFB3 enzymatic activity in HR repair and present KAN0438757, a selective PFKFB3 inhibitor that could potentially be used as a strategy for the treatment of cancer.
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Sulforhodamine B based
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-RRM2 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade