Merck
  • Guanidinium ionic liquid-based surfactants as low cytotoxic extractants: Analytical performance in an in-situ dispersive liquid-liquid microextraction method for determining personal care products.
Guanidinium ionic liquid-based surfactants as low cytotoxic extractants: Analytical performance in an in-situ dispersive liquid-liquid microextraction method for determining personal care products.
Journal of chromatography. A (2017-05-11)
Idaira Pacheco-Fernández, Verónica Pino, Juan H Ayala, Ana M Afonso
ABSTRACT
The IL-based surfactant octylguanidinium chloride (C8Gu-Cl) was designed and synthetized with the purpose of obtaining a less harmful surfactant: containing guanidinium as core cation and a relatively short alkyl chain. Its interfacial and aggregation behavior was evaluated through conductivity and fluorescence measurements, presenting a critical micelle concentration value of 42.5 and 44.6mmolL-1, respectively. Cytotoxicity studies were carried out with C8Gu-Cl and other IL-based and conventional surfactants, specifically the analogue 1-octyl-3-methylimidazolium chloride (C8MIm-Cl), and other imidazolium- (C16MIm-Br) and pyridinium- (C16Py-Cl) based surfactants, together with the conventional cationic CTAB and the conventional anionic SDS. From these studies, C8Gu-Cl was the only one to achieve the classification of low cytotoxicity. An in situ dispersive liquid-liquid microextraction (DLLME) method based on transforming the water-soluble C8Gu-Cl IL-based surfactant into a water-insoluble IL microdroplet via a simple metathesis reaction was then selected as the extraction/preconcentration method for a group of 6 personal care products (PCPs) present in cosmetic samples. The method was carried out in combination with high-performance liquid chromatography (HPLC) and diode array detection (DAD). The method was properly optimized, requiring the use of only 30μL of C8Gu-Cl for 10mL of aqueous sample with a NaCl content of 8% (w/v) to adjust the ionic strength and pH value of 5. The metathesis reaction required the addition of the anion exchange reagent (bis[(trifluoromethyl)sulfonyl]imide - 1:1 molar ratio), followed by vortex and centrifugation, and dilution of the final microdroplet up to 60μL with acetonitrile before the injection in the HPLC-DAD system. The optimum in situ DLLME-HPLC-DAD method takes ∼10min for the extraction step and ∼22min for the chromatographic separation, with analytical features of low detection limits: down to 0.4μgL-1; high reproducibility: with RSD values lower than 10% (intra-day) and 16% (inter-day) for a spiked level of 15μgL-1; and an average enrichment factor of 89. The requirement of low volumes (30μL) of a low cytotoxic IL-based surfactant allows the method to be considered less harmful than other common analytical microextraction approaches.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
1-Methyl-3-octylimidazolium chloride, ≥97.0% (HPLC)
Sigma-Aldrich
Octylamine, ≥99.5%