MilliporeSigma
p62 filaments capture and present ubiquitinated cargos for autophagy.
The EMBO journal (2018-01-19)
Gabriele Zaffagnini, Adriana Savova, Alberto Danieli, Julia Romanov, Shirley Tremel, Michael Ebner, Thomas Peterbauer, Martin Sztacho, Riccardo Trapannone, Abul K Tarafder, Carsten Sachse, Sascha Martens
ABSTRACT
The removal of misfolded, ubiquitinated proteins is an essential part of the protein quality control. The ubiquitin-proteasome system (UPS) and autophagy are two interconnected pathways that mediate the degradation of such proteins. During autophagy, ubiquitinated proteins are clustered in a p62-dependent manner and are subsequently engulfed by autophagosomes. However, the nature of the protein substrates targeted for autophagy is unclear. Here, we developed a reconstituted system using purified components and show that p62 and ubiquitinated proteins spontaneously coalesce into larger clusters. Efficient cluster formation requires substrates modified with at least two ubiquitin chains longer than three moieties and is based on p62 filaments cross-linked by the substrates. The reaction is inhibited by free ubiquitin, K48-, and K63-linked ubiquitin chains, as well as by the autophagosomal marker LC3B, suggesting a tight cross talk with general proteostasis and autophagosome formation. Our study provides mechanistic insights on how substrates are channeled into autophagy.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Glutathione-S-transferase (GST) antibody, Mouse monoclonal, clone 2H3-D10, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)