Skip to Content
Merck
vibrant-m

Ziprasidone Related Compound F

vibrant-m
Synonym(s):
Sodium 2-(2-amino-5-{2-[4-(benzo[d]isothiazol-3-yl)piperazin-1-yl]ethyl}-4-chlorophenyl)acetate monohydrate
Empirical Formula (Hill Notation):
C21H22ClN4NaO2S · H2O
Molecular Weight:
470.95

Products

Formula Weight
Manufacturer
Physical Form
Application
API Family
Reference Material Type
Showing 1-1 of 1