Skip to Content
Merck
Tetramethylammonium fluoride tetrahydrate 98%

Soy Peptone for Biotechnology and Microbiology

Tetramethylammonium fluoride tetrahydrate 98%
Linear Formula:
(CH3)4N(F) · 4H2O
CAS Number:
Molecular Weight:
165.20

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group