Skip to Content
Merck
Sodium triflate ≥99.5% trace metals basis, anhydrous, battery grade

Sodium triflate

Sodium triflate ≥99.5% trace metals basis, anhydrous, battery grade
Synonym(s):
Sodium trifluoromethanesulfonate, Sodium triflate, Trifluoromethanesulfonic acid sodium salt
Linear Formula:
CF3SO3Na
CAS Number:
Molecular Weight:
172.06

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing