Skip to Content
Merck
Peptone from pea suitable for microbiology

Pea Peptone for Biotechnology and Microbiology

Peptone from pea suitable for microbiology
Synonym(s):
Peptone from vegetable
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing