
Olsalazine sodium for performance test

Synonym(s):
4,4′-Dihydroxyazobenzene-3,3′-dicarboxylic acid disodium salt, 5,5′-Azobis(salicylic acid, sodium salt), Mordant Yellow 5
Empirical Formula (Hill Notation):
C14H8N2Na2O6
CAS Number:
Molecular Weight:
346.20
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing