
N-Nitroso-N,N-bis(8-methylnonyl)amine

Empirical Formula (Hill Notation):
C20H42N2O
Molecular Weight:
326.56
Products
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing