
Mesoridazine besylate

Synonym(s):
Mesoridazine benzenesulfonate, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine
Empirical Formula (Hill Notation):
C27H32N2O4S3
CAS Number:
Molecular Weight:
544.75
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing