
Mefenamic acid solution

Synonym(s):
Mefenamic acid, 2-[(2,3-Dimethylphenyl)amino]benzoic acid, N-(2,3-Xylyl)anthranilic acid
Empirical Formula (Hill Notation):
C15H15NO2
CAS Number:
Molecular Weight:
241.29
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing