Skip to Content
Merck
Lipopolysaccharides (rough strains) from Escherichia coli J5 (Rc mutant)

Lipopolysaccharides (rough strains) from Escherichia coli J5 (Rc mutant)

Lipopolysaccharides (rough strains) from Escherichia coli J5 (Rc mutant)
Synonym(s):
LPS
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Color
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing