Skip to Content
Merck
Lipopolysaccharides from Escherichia coli O26:B6 purified by trichloroacetic acid extraction

Lipopolysaccharides from Escherichia coli O26:B6

Lipopolysaccharides from Escherichia coli O26:B6 purified by trichloroacetic acid extraction
Synonym(s):
LPS
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique