
Fluazifop-methyl

Synonym(s):
Fluazifop-methyl, Methyl 2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate
Empirical Formula (Hill Notation):
C16H14F3NO4
CAS Number:
Molecular Weight:
341.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing