Skip to Content
Merck
Ethanol standards 10% (v/v) 10 % (v/v) in H2O, analytical standard

Ethanol standards 10% (v/v)

Ethanol standards 10% (v/v) 10 % (v/v) in H2O, analytical standard
Synonym(s):
Ethanol solution, Ethyl alcohol
Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07

Products

Product Category
Formula Weight
Format
Quality Segment
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing