
Estradiol Related Compound B

Synonym(s):
1,3,5(10),6-Estratetraen-3,17β-diol, 6-Dehydroestradiol
Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing