Skip to Content
Merck
vibrant-m

Dipyridamole Related Compound A

vibrant-m
Synonym(s):
2,2′-[(4,6,8-Tri-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl)imino]bisethanol, 2,2′-{[4,6,8-Tri(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]azanediyl}diethanol
Empirical Formula (Hill Notation):
C25H40N8O2
CAS Number:
Molecular Weight:
484.64

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1