
cis-Dichloroethylene

Synonym(s):
cis-1,2-Dichloroethylene, cis-Acetylene dichloride
Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing