Skip to Content
Merck
vibrant-m

Budesonide Related Compound E

vibrant-m
Synonym(s):
16α,17-[Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4,14-triene-3,20-dione
Empirical Formula (Hill Notation):
C25H32O6
CAS Number:
Molecular Weight:
428.52

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1