
Brinzolamide Related Compound B

Synonym(s):
(R)-4-Amino-3,4-dihydro-2-(3-methoxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-dioxide ethandioate
Empirical Formula (Hill Notation):
C10H17N3O5S3 · C2H2O4
CAS Number:
Molecular Weight:
445.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing