Skip to Content
Merck
Anti-SNW1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-SNW1 antibody produced in rabbit

Anti-SNW1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Nuclear protein SkiP antibody produced in rabbit, Anti-Nuclear receptor coactivator NCoA-62 antibody produced in rabbit, Anti-SNW domain-containing protein 1 antibody produced in rabbit, Anti-Ski-interacting protein antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique