
Anti-MYH-1 Antibody

Synonym(s):
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, Anti-MGC133384, Anti-MYHSA1, Anti-MYHa, Anti-MyHC-2X/D
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing