Skip to Content
Merck
MONOCLONAL ANTI-ERK1/2 antibody produced in mouse clone 784CT7.6.3, IgG fraction of antiserum, buffered aqueous solution

Anti-MAP Kinase- ERK1/ERK2 Antibody

MONOCLONAL ANTI-ERK1/2 antibody produced in mouse clone 784CT7.6.3, IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-MAP Kinase (ERK-1, ERK-2), Anti-Mitogen Activated Protein Kinase, ERK1, MAPK3, Mitogen-activated protein kinase 3, PRKM3

Products

Product Category
Brand
Biological Source
Clonality
Color
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Color
Package Size
Post Translational Modification
Species Reactivity
Technique