
Anti-MAPK3/ERK1 Antibody

Synonym(s):
Anti-ERK-1 antibody produced in rabbit, Anti-ERT2 antibody produced in rabbit, Anti-Extracellular signal-regulated kinase 1 antibody produced in rabbit, Anti-Insulin-stimulated MAP2 kinase antibody produced in rabbit, Anti-MAP kinase 1 antibody produced in rabbit, Anti-MAPK 1 antibody produced in rabbit, Anti-Microtubule-associated protein 2 kinase antibody produced in rabbit, Anti-Mitogen-activated protein kinase 3 antibody produced in rabbit, Anti-p44-ERK1 antibody produced in rabbit, Anti-p44-MAPK antibody produced in rabbit, ERK-1, ERK1, ERT2, Extracellular signal- regulated kinase 1, Insulin-stimulated MAP2 kinase, extracellular signal-regulated kinase 1, insulin-stimulated MAP2 kinase, Anti-ERK1 antibody produced in rabbit, Anti-HS44KDAP antibody produced in rabbit, Anti-HUMKER1A antibody produced in rabbit, Anti-MGC20180 antibody produced in rabbit, Anti-mitogen-activated protein kinase 3 antibody produced in rabbit, Anti-ERK1, Anti-P44ERK1, Anti-P44MAPK, Anti-PRKM3, Anti-mitogen-activated protein kinase 3, HS44KDAP, P44ERK1, P44MAPK, PRKM3, 3pK, Chromosome 3p kinase, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAPK-3, HUMKER1A, MAPK3, MGC20180
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing