Skip to Content
Merck
3′-(N,N-Didemethyl)Azithromycin; (Aminoazithromycin) Pharmaceutical Analytical Impurity (PAI)

3′-(N,N-Didemethyl)Azithromycin; (Aminoazithromycin)

3′-(N,N-Didemethyl)Azithromycin; (Aminoazithromycin) Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
((2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-amino-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2- ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)- 3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3-amino-3,4,6-trideoxy-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6- azacyclopentadecan-15-one)
Empirical Formula (Hill Notation):
C36H68N2O12
CAS Number:
Molecular Weight:
720.93

Products

Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1