![2-Methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/235/582/d6e10a33-082f-4c26-bf03-811df8e5f3fe/640/d6e10a33-082f-4c26-bf03-811df8e5f3fe.jpg)
2-Methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide
![2-Methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/235/582/d6e10a33-082f-4c26-bf03-811df8e5f3fe/640/d6e10a33-082f-4c26-bf03-811df8e5f3fe.jpg)
Empirical Formula (Hill Notation):
C18H24N4O
CAS Number:
Molecular Weight:
312.41
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1