
2,5-Deoxyfructosazine

Synonym(s):
(1R,2S,3R)-1-[5-[(2S,3R)-2,3,4-Trihydroxybutyl]-2-pyrazinyl]-1,2,3,4-butanetetrol
Empirical Formula (Hill Notation):
C12H20N2O7
CAS Number:
Molecular Weight:
304.30
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing