Product Name
Progranulin mouse recombinant, expressed in HEK 293 cells, FLAG® tagged, >95% (SDS-PAGE)
Product Number
SRP8016
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Source | recombinant expressed in HEK 293 cells |
Molecular Weight | ~70 kDa (SDS-PAGE) |
Storage/Stability | -20�C. Working aliquots are stable for up to 3 months when stored at -20�C. Avoid repeated freeze/thaw cycles. |
Formulation | Solution in PBS. |
Form | liquid |
Color | clear solution |
Concentration | >=0.2 mg/ml |
Purity by SDS-PAGE | >=95% |
Endotoxin Level | <0.1 EU/ug purified protein (LAL test). |