Product Name
p75 human recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Product Number
SRP2026
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | frozen liquid solution |
Color | colorless and clear |
Concentration | 500 ug/mL |
Purity by SDS Page | >=70 % |