Product Name
Anti-Phospho-MAPKAPK2-T334 antibody produced in rabbit
Product Number
SAB5701778
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |