Product Name
ANTI-TFAP4 antibody produced in mouse clone 8G6, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1412283
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | Report Result |
Storage/Stability | Report Result |
Storage Temperature | -20�C |
Species Reactivity | Report Result |
ELISA | Antibody concentration: 1 ug/ml using recombinant protein. |
Sandwich ELISA | Detection limit for recombinant GST tagged TFAP4 is approximately 0.1ng/ml. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1 ug/ml. Specific band of ~36.7 kDa using immunogen protein lysate. |
Western Blotting (Cell Lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~39 kDa using Jurkat lysate. |
Western Blotting (Tissue lysate) | Not Verified |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~38.9 kDa using TFAP4 transfected 293T lysate. |
Immunohistochemistry | Antibody concentration: 1.5 ug/ml using human small Intestine tissue. |
Immunofluorescence | Not Verified |
Handling | Report Result |