Product Name
ANTI-TOP3B antibody produced in mouse clone 2G2, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1412102
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | Report Result |
Storage/Stability | Report Result |
Storage Temperature | -20�C |
Species Reactivity | Report Result |
ELISA | Antibody concentration: 1 ug/ml using recombinant protein. |
Sandwich ELISA | Not Verified |
Western Blotting (Recombinant Protein) | Antibody concentration: 1 ug/ml. Specific band of ~120.6 kDa using immunogen protein lysate. |
Western Blotting (Cell Lysate) | Not Verified |
Western Blotting (Tissue lysate) | Not Verified |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~96.7 kDa using TOP3B transfected 293T lysate. |
Immunohistochemistry | Not Verified |
Immunofluorescence | Not Verified |
Handling | Report Result |