Product Name
Anti-Cannabinoid Receptor 1 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
C2866
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Formulation | Conforms |
Solution in phosphate buffered saline, pH 7.7, | |
containing 0.1% sodium azide | |
Concentration | Conforms |
1 mg/mL | |
Working Dilution by Immunohistochemistry | Conforms |
8-11 ug/mL using brain and neurons tissues |