Product Name
Atto 425 NHS ester BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
16805
Product Brand
SIGMA
CAS Number
892156-28-4
Molecular Weight
498.53
Test
Specification
APPEARANCE (COLOR)Report Result
APPEARANCE (FORM)Report Result
PURITY (HPLC AREA %)≥ 90 %
Absorption Max422 - 428 nm
SOLVENT (UV)ethanol with 0.1% trifluoroacetic acid
DEGREE OF COUPLING≥ 90 %
RECOMMENDED RETEST PERIOD28 MONTHS