Product Name
Tetrahydrofuran contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Product Number
87368
Product Brand
SIGALD
CAS Number
109-99-9
Molecular Weight
72.11
Test | Specification |
---|---|
STABILIZER | STABILIZED WITH ~0.025 % 2,6-DI-TERT-BUTYL-4-METHYLPHENOL |
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
COLOR (IN APHA) | ≤ 20 APHA |
PURITY (GC AREA %) | ≥ 99.9 % |
DENSITY D20/4 | 0.886 - 0.888 |
REFRACTIVE INDEX N20/D | 1.406 - 1.408 |
WATER (COULOMETR.) | ≤ 0.05 % |
RESIDUE (EVAPORATION) | ≤ 0.001 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
MISCELLANEOUS TESTS | MISCIBLE WITH WATER: PASS; MISCIBLE WITH ETHANOL (96 %): PASS |
TRACES PEROXIDE (AS H2O2) | ≤ 50 mg/kg |
TRACE ANALYSIS (ADDITIONAL TESTS) | FREE ACID (AS CH3COOH) ≤ 0.001 % |
ACS SPECIFICATIONS | CORRESPONDS TO REQUIREMENTS |
ALUMINIUM (ICP) | ≤ 0.5 mg/kg |
BARIUM (ICP) | ≤ 0.1 mg/kg |
BISMUTH (ICP) | ≤ 0.1 mg/kg |
BORON (ICP) | ≤ 0.02 mg/kg |
CALCIUM (ICP) | ≤ 0.5 mg/kg |
CADMIUM (ICP) | ≤ 0.05 mg/kg |
COBALT (ICP) | ≤ 0.02 mg/kg |
CHROMIUM (ICP) | ≤ 0.02 mg/kg |
COPPER (ICP) | ≤ 0.02 mg/kg |
IRON (ICP) | ≤ 0.1 mg/kg |
POTASSIUM (ICP) | ≤ 0.5 mg/kg |
LITHIUM (ICP) | ≤ 0.1 mg/kg |
MAGNESIUM (ICP) | ≤ 0.1 mg/kg |
MANGANESE (ICP) | ≤ 0.02 mg/kg |
MOLYBDENUM (ICP) | ≤ 0.1 mg/kg |
SODIUM (ICP) | ≤ 0.5 mg/kg |
NICKEL (ICP) | ≤ 0.02 mg/kg |
LEAD (ICP) | ≤ 0.1 mg/kg |
TIN (ICP) | ≤ 0.1 mg/kg |
STRONTIUM (ICP) | ≤ 0.1 mg/kg |
ZINC (ICP) | ≤ 0.1 mg/kg |
METAL TRACE ANALYSIS (ICP-MS) | CORRESPONDS TO REQUIREMENTS |
RECOMMENDED RETEST PERIOD | 42 MONTHS |