Product Name
D-(+)-Raffinose pentahydrate suitable for microbiology, ≥99.0%, Microbiological media component
Product Number
83400
Product Brand
SIAL
CAS Number
17629-30-0
Molecular Weight
594.51
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 99.0 % |
SPECIFIC ROTATION (20/D) | 105.0 ± 2.0 Degrees |
CONCENTRATION | C=10 IN WATER |
MELTING POINT | 78 - 80 C |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1G IN 10ML WATER, HOT |
WATER | 13.0 - 17.0 % |
SULFATED ASH | ≤ 0.1 % |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
CALCIUM (ICP) | ≤ 20 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 5 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 5 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
CHLORIDE (CL) | ≤ 50 mg/kg |