Product Name
Butylated hydroxyanisole analytical standard
Product Number
78943
Product Brand
SIAL
CAS Number
25013-16-5
Molecular Weight
180.24
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige |
APPEARANCE (FORM) | Powder to Crystals |
PURITY (GC AREA %) | ≥ 99.0% (sum of isomers) |
WATER | ≤ 0.3 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |