Product Name
Sodium phosphate dibasic purum p.a., anhydrous, ≥98.0% (T)
Product Number
71642
Product Brand
SIAL
CAS Number
7558-79-4
Molecular Weight
141.96
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
TITRATION | 98.0 - 102.0 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 25C water 50mg/mL |
PH | 8.8 - 9.4 |
LOSS ON DRYING | ≤ 2 % 110 C |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
CALCIUM (ICP) | ≤ 50 mg/kg |
CADMIUM (ICP) | ≤ 50 mg/kg |
COBALT (ICP) | ≤ 50 mg/kg |
COPPER (ICP) | ≤ 50 mg/kg |
IRON (ICP) | ≤ 50 mg/kg |
POTASSIUM (ICP) | ≤ 200 mg/kg |
NICKEL (ICP) | ≤ 50 mg/kg |
LEAD (ICP) | ≤ 50 mg/kg |
ZINC (ICP) | ≤ 50 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 1000 mg/kg |
CHLORIDE (CL) | ≤ 50 mg/kg |
RECOMMENDED RETEST PERIOD | 36 MONTHS |