Product Name
Dihexyl phthalate analytical standard
Product Number
49617
Product Brand
SIAL
CAS Number
84-75-3
Molecular Weight
334.45
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
REFRACTIVE INDEX N20/D | 1.486 - 1.489 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |