Product Name
3,4-Pyridinedicarboxylic acid 97%
Product Number
P64006
Product Brand
ALDRICH
CAS Number
490-11-9
Molecular Weight
167.12
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER OR CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH NAOH) |
THIN-LAYER | CONSISTENT WITH 97% PURITY |
SOLUBILITY | 5%, 1N NH4OH; CLEAR TO HAZY, COLORLESS TO |
YELLOW SOLUTION | |
APPROVED NOVEMBER 19, 2008 RJM |