Product Name
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone 99%
Product Number
458775
Product Brand
ALDRICH
CAS Number
101711-78-8
Molecular Weight
233.26
Test | Specification |
---|---|
APPEARANCE | WHITE SOLID, POWDER OR CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 65.9%-67.9% |
NITROGEN 5.8%- 6.2% | |
OPTICAL ROTATION | +101(+/-5) DEGREES (C=1%, ETHANOL) |
ENANTIOMERIC EXCESS | 98.5% (MINIMUM) |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED OCTOBER 10, 2008 RJM |