Product Name
1,3-Dimethyluracil 99%
Product Number
349801
Product Brand
ALDRICH
CAS Number
874-14-6
Molecular Weight
140.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | WHITE TO LIGHT YELLOW OR TAN |
APPEARANCE (FORM) | Powder |
PURITY (GC AREA %) | ≥ 98.5 % |
CARBON CONTENT | 50.7 - 52.2 % |
NITROGEN CONTENT | 19.7 - 20.3 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |