Product Name
Bis(2-ethylhexyl) sebacate technical grade, 90%
Product Number
290831
Product Brand
ALDRICH
CAS Number
122-62-3
Molecular Weight
426.67
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 89.0% (MINIMUM) |
REVISED JUNE 10, 2005 RJM |