Product Name
Ethyl acetimidate hydrochloride 97%
Product Number
188840
Product Brand
ALDRICH
CAS Number
2208-07-3
Molecular Weight
123.58
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER, CRYSTALS, CRYSTALLINE POWDER |
AND/OR CHUNKS | |
IDENTITY | CONFORMS TO STRUCTURE BY IR OR HNMR. |
TITRATION | 96.5%-103.5% (WITH SILVER NITRATE) |
SOLUBILITY | 50MG/ML(5%)H2O; CLEAR, COLORLESS SOLUTION |
APPROVED OCTOBER 2006 RJM |