Product Name
Cyanoacetamide 99%
Product Number
108448
Product Brand
ALDRICH
CAS Number
107-91-5
Molecular Weight
84.08
Test | Specification |
---|---|
Appearance (Color) | Conforms to Requirements |
White to Off-White or Yellow | |
Appearance (Form) | Conforms to Requirement |
Powder and/or Chunks | |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥98.5 % |